missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Iron(II) bromide hydrate

Manufacturer:  Alfa Aesar™ 3113322

 View more versions of this product

Catalog No. AA3113322



Specifications

Specifications

Iron(II) bromide hydrate
Br2Fe
MFCD00016081
iron dibromide, unii-ea3x054rbz, iron ii bromide, ea3x054rbz, iron 2+ dibromide, iron ion 2+ dibromide, iron ii bromide, anhydrous 5g, iron ii bromide, ultra dry metals basis, ferrous bromide, anhydrous
[Fe+2].[Br-].[Br-]
215.653
215.67 (Anhydrous)
3.9g/mL
Air Sensitive & Hygroscopic
27°C
7789-46-0
FeBr2·xH2O
UN3260
GYCHYNMREWYSKH-UHFFFAOYSA-L
iron(2+);dibromide
82240
Lumps
Odorless
Green
100g

Chemical Identifiers

7789-46-0
215.653
GYCHYNMREWYSKH-UHFFFAOYSA-L
82240
[Fe+2].[Br-].[Br-]
Br2Fe
MFCD00016081
iron dibromide, unii-ea3x054rbz, iron ii bromide, ea3x054rbz, iron 2+ dibromide, iron ion 2+ dibromide, iron ii bromide, anhydrous 5g, iron ii bromide, ultra dry metals basis, ferrous bromide, anhydrous
iron(2+);dibromide
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318-H302
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.

GHS P Statement
P280-P305+P351+P338-P309-P310
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, INORGANIC, N.O.S.

TSCA : TSCA

SDS Documents