Please login to your online account to display your discounted pricing

Iron(II) acetate, 95%, ACROS Organics™


Quantity 250g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC305372500

 View more versions of this product

Catalog No. AC305372500

  • / Each
 

Description & Specifications

Chemical Identifiers

Physical Form Powder
Assay ≥94% (with KMnO4)
Formula Weight 173.93
Quantity 250g
Color Off-white to light gray or light brown
TSCA TSCA
RTECS Number AI3850000
EINECS Number 221-441-7
Infrared Spectrum Authentic
Packaging Glass bottle
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 1 Warning
Chemical Name or Material Iron(II) acetate
Linear Formula (CH3CO2)2
Assay Percent Range 95%
Name Note 95%
Molecular Formula C4H6O4·Fe
CAS 3094-87-9
MDL Number MFCD00058909
Synonym Ferrous acetate
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

Specifications

Chemical Name or Material Iron(II) acetate
Name Note 95%
CAS 3094-87-9
Assay ≥94% (with KMnO4)
Assay Percent Range 95%
Molecular Formula C4H6O4·Fe
Linear Formula (CH3CO2)2
Formula Weight 173.93
MDL Number MFCD00058909
Synonym Ferrous acetate
Accessories

Warning

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

SDS & Safety Documents