Please login to your online account to display your discounted pricing

Alfa Aesar™ Iron(II) 2-ethylhexanoate monoisopropoxide, 5% w/v in hexane

Chemical Identifiers

Chemical Name or Material Iron(II) 2-ethylhexanoate monoisopropoxide
CAS 331686-36-3
Formula Weight 258.14
MDL Number MFCD00798541
UN Number UN1208
Synonym Iron(II) 2-ethylhexano-isopropoxide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA40584AC

  SDS
alfa aesar™
40584AC
25mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA40584AE

  SDS
alfa aesar™
40584AE
100mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 258.14
Chemical Name or Material Iron(II) 2-ethylhexanoate monoisopropoxide
Synonym Iron(II) 2-ethylhexano-isopropoxide
Health Hazard 2 GHS H Statement
H225-H304-H361-H373-H315-H319-H336
Highly flammable liquid and vapor.
May be fatal if swallowed and enters airways.
Suspected of damaging fertility or the unborn child.
May cause damage to organs through prolonged or repeated exposure.
Causes skin irritation.
Causes serious eye irritation.
May cause drowsiness or dizziness.
Quantity 25mL
MDL Number MFCD00798541
Health Hazard 3 GHS P Statement
P210-P280h-P273-P305+P351+P338-P301+P330+P331-P315
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Wear protective gloves/clothing.
Avoid release to the environment.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Get immediate medical advice/attention.
DOT Information Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: HEXANES
CAS 331686-36-3
Concentration 5% w/v in hexane
Health Hazard 1 Danger
Physical Form Liquid
UN Number UN1208
Sensitivity Moisture Sensitive
Safety and Handling

Danger

GHS P Statement
P210-P280h-P273-P305+P351+P338-P301+P330+P331-P315
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Wear protective gloves/clothing.
Avoid release to the environment.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Get immediate medical advice/attention.

GHS H Statement
H225-H304-H361-H373-H315-H319-H336
Highly flammable liquid and vapor.
May be fatal if swallowed and enters airways.
Suspected of damaging fertility or the unborn child.
May cause damage to organs through prolonged or repeated exposure.
Causes skin irritation.
Causes serious eye irritation.
May cause drowsiness or dizziness.