missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Iron ICP Standard, 1mL = 10mg Fe (10,000ppm Fe), Ricca Chemical

Manufacturer:  Ricca Chemical Company RIPFE10KN50

 View more versions of this product

Catalog No. PFE10KN-50

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

Iron ICP Standard
84.0,12.69,0.85
Fe
[Fe]
55.845
CHEBI:82664
Poly natural
1 mL = 10 mg Fe
50mL
7439-89-6,7697-37-2,7732-18-5
88.0,13.21,0.88
XEEYBQQBJWHFJM-UHFFFAOYSA-N
iron
23925
Liquid
10,000ppm Fe
Colorless to Yellow

Chemical Identifiers

7439-89-6
55.845
23925
Fe
XEEYBQQBJWHFJM-UHFFFAOYSA-N
CHEBI:82664
Documents