Please login to your online account to display your discounted pricing

Iron ICP Standard, 1mL = 10mg Fe (10,000ppm Fe), Ricca Chemical

$213.26 - $213.26

Chemical Identifiers

Chemical Name or Material Iron ICP Standard
CAS 7732-18-5
CAS Min % 84.0
CAS Max % 88.0
CAS 7697-37-2
CAS Min % 12.69
CAS Max % 13.21
CAS 7439-89-6
CAS Min % 0.85
CAS Max % 0.88
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

PFE10KN-50

 
ricca chemical company
RIPFE10KN50
Poly natural 50mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

PFE10KN-500

 
ricca chemical company
RIPFE10KN500
Poly natural 500mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

PFE10KN-100

 
ricca chemical company
RIPFE10KN100
Poly natural 100mL Each for $213.26
Description & Specifications

Specifications

Physical Form Liquid
Color Clear, colorless to faint yellow
Quantity 50mL
Chemical Name or Material Iron ICP Standard
CAS 7732-18-5
Equivalence 1 mL = 10 mg Fe
Concentration or Composition Notes 10,000ppm Fe
CAS Min % 0.85
CAS Min % 84.0
CAS Max % 13.21
CAS Max % 0.88
CAS 7439-89-6
Packaging Poly natural
CAS Min % 12.69
CAS Max % 88.0
CAS 7697-37-2