missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Iron, For Analysis Reduced, Particle Size 10um EMSURE™, MilliporeSigma™

Manufacturer:  MilliporeSigma 1038190500

 View more versions of this product

Catalog No. M1038190500



Specifications

Specifications

Iron
7439-89-6
Fe
ferrum, iron, elemental, ferrous, remko, armco, ferrovac, hoeganaes eh, 3zhp, ancor b, atomiron 5m
[Fe]
Odorless
Plastic bottle
CHEBI:82664
Solid
500g
Gray solid
UN 3089
XEEYBQQBJWHFJM-UHFFFAOYSA-N
iron
55.845
23925
55.85

Chemical Identifiers

7439-89-6
55.845
ferrum, iron, elemental, ferrous, remko, armco, ferrovac, hoeganaes eh, 3zhp, ancor b, atomiron 5m
CHEBI:82664
[Fe]
Fe
XEEYBQQBJWHFJM-UHFFFAOYSA-N
23925
iron
Safety and Handling

Safety and Handling

H228: Flammable solid

DOTInformation : DOT Class: 4.1, Packaging Group: III

EINECSNumber : 231-096-4

Documents
Promotions

Promotions