missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Iron foil, 99.995% (metals basis), Alfa Aesar™, Puratronic™

Form: approx. 0.49g/25x25mm

Manufacturer:  Alfa Aesar 11381FF

 View more versions of this product

Catalog No. AA11381FF



Specifications

Specifications

Iron
7439-89-6
99.995% (Metals basis)
MFCD00010999
XEEYBQQBJWHFJM-UHFFFAOYSA-N
iron
23925
Foil, 0.1mm (0.004 in.) thick
25 x 25mm
Gray foil
Fe
ferrum, iron, elemental, ferrous, remko, armco, ferrovac, hoeganaes eh, 3zhp, ancor b, atomiron 5m
[Fe]
55.845
CHEBI:82664
Gray

Chemical Identifiers

7439-89-6
55.845
XEEYBQQBJWHFJM-UHFFFAOYSA-N
23925
iron
Fe
MFCD00010999
ferrum, iron, elemental, ferrous, remko, armco, ferrovac, hoeganaes eh, 3zhp, ancor b, atomiron 5m
CHEBI:82664
[Fe]
Safety and Handling

Safety and Handling

EINECSNumber : 231-096-4

Documents