missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Iron foil, 1.0mm (0.04 in.) thick, Puratronic™, 99.995% (metals basis)

Manufacturer:  Alfa Aesar™ 11448GH

 View more versions of this product

Catalog No. AA11448GH



Specifications

Specifications

Iron foil
100 x 100mm
7439-89-6
Fe
3zhp
[Fe]
55.845
CHEBI:82664
99.995%
≈4.9g/25x25mm
1.0mm (0.04 in.) thick
(metals basis)
MFCD00010999
XEEYBQQBJWHFJM-UHFFFAOYSA-N
iron
23925
Foil
Puratronic™

Chemical Identifiers

7439-89-6
55.845
XEEYBQQBJWHFJM-UHFFFAOYSA-N
23925
iron
Fe
MFCD00010999
3zhp
CHEBI:82664
[Fe]
Safety and Handling

Safety and Handling

TSCA : TSCA

Documents