missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Iron foil, 0.5mm (0.02 in.) thick, Puratronic™, 99.994% (metals basis)

Manufacturer:  Alfa Aesar™ 11379FF

 View more versions of this product

Catalog No. AA11379FF



Specifications

Specifications

Iron foil
25 x 25mm
7439-89-6
Fe
3zhp
[Fe]
55.845
CHEBI:82664
99.994%
≈2.5g/25x25mm
0.5mm (0.02 in.) thick
(metals basis)
MFCD00010999
XEEYBQQBJWHFJM-UHFFFAOYSA-N
iron
23925
Foil
Puratronic™

Chemical Identifiers

7439-89-6
55.845
XEEYBQQBJWHFJM-UHFFFAOYSA-N
23925
iron
Fe
MFCD00010999
3zhp
CHEBI:82664
[Fe]
Safety and Handling

Safety and Handling

TSCA : TSCA

Documents