missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Iron (Electrolytic Powder), Fisher Chemical

$386.08 - $386.08

Chemical Identifiers

CAS 7439-89-6
Molecular Formula Fe
Molecular Weight (g/mol) 55.845
MDL Number MFCD00010999
InChI Key XEEYBQQBJWHFJM-UHFFFAOYSA-N
Synonym 3zhp, ancor b, armco, atomiron 5m, ferrous, ferrovac, ferrum, hoeganaes eh, iron, elemental, remko
PubChem CID 23925
ChEBI CHEBI:82664
IUPAC Name iron
SMILES [Fe]
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
I60-500 SDS View Documents Fisher Chemical
I60500
500g Glass Bottle Each for $386.08
Case of 6 Each for $2,316.48
 
Specifications

Chemical Identifiers

7439-89-6
55.845
XEEYBQQBJWHFJM-UHFFFAOYSA-N
23925
iron
Fe
MFCD00010999
3zhp, ancor b, armco, atomiron 5m, ferrous, ferrovac, ferrum, hoeganaes eh, iron, elemental, remko
CHEBI:82664
[Fe]

Specifications

Vapor Pressure: 1mmHg at 1787°C
7439-89-6
Pass test
Fe
3089
3zhp, ancor b, armco, atomiron 5m, ferrous, ferrovac, ferrum, hoeganaes eh, iron, elemental, remko
[Fe]
55.845
23925
55.85
≥99%
Gray
3000°C
500g
7.86g/mL
≥99 %
MFCD00010999
Finer than 100 Mesh
XEEYBQQBJWHFJM-UHFFFAOYSA-N
iron
Glass Bottle
CHEBI:82664
Powder/Solid
55.85
1535°C
Iron
Safety and Handling

Safety and Handling

DOTInformation : DOT Class 4.1, : Flammable Solid