Please login to your online account to display your discounted pricing

Alfa Aesar™ Iron disilicide, 99.9% (metals basis)

$93.42 - $93.42

Chemical Identifiers

Chemical Name or Material Iron disilicide
CAS 12022-99-0
Formula Weight 112.02
MDL Number MFCD00016087
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA1402214

  SDS
alfa aesar™
1402214
25g Each for $93.42

AA1402206

  SDS
alfa aesar™
1402206
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 112.02
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Odor Odorless
Assay 99.9%
Chemical Name or Material Iron disilicide
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Quantity 25g
MDL Number MFCD00016087
CAS 12022-99-0
Health Hazard 1 Warning
Melting Point 1220°C
Assay Percent Notes (metals basis)
Mesh Size −20 Mesh
TSCA TSCA
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.