missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Iron Dextran, Fe 33-40% w/w

Prevents and treats neonatal anemia in pigs

$83.07 - $291.31

Chemical Identifiers

CAS 9004-66-4
Molecular Formula FeH2O4S
Molecular Weight (g/mol) 153.917
MDL Number MFCD00081553
InChI Key MVZXTUSAYBWAAM-UHFFFAOYSA-N
Synonym conferon, fero gradumet, ferridextran, iron dextran, iron-dextran, iron; sulfuric acid, sulfuric acid iron 2+ salt 1:1, sulfuric acid iron salt 1:1, sulfuric acid, iron salt, sulphuric acid, iron salt
PubChem CID 105075
IUPAC Name iron;sulfuric acid
SMILES OS(=O)(=O)O.[Fe]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAJ6275030 View Documents Alfa Aesar™
J6275030
250g Each for $83.07
 
AAJ62750A1 View Documents Alfa Aesar™
J62750A1
1kg Each for $291.31
 
Specifications

Chemical Identifiers

9004-66-4
153.917
MVZXTUSAYBWAAM-UHFFFAOYSA-N
105075
OS(=O)(=O)O.[Fe]
FeH2O4S
MFCD00081553
conferon, fero gradumet, ferridextran, iron dextran, iron-dextran, iron; sulfuric acid, sulfuric acid iron 2+ salt 1:1, sulfuric acid iron salt 1:1, sulfuric acid, iron salt, sulphuric acid, iron salt
iron;sulfuric acid

Specifications

9004-66-4
MFCD00081553
conferon, fero gradumet, ferridextran, iron dextran, iron-dextran, iron; sulfuric acid, sulfuric acid iron 2+ salt 1:1, sulfuric acid iron salt 1:1, sulfuric acid, iron salt, sulphuric acid, iron salt
OS(=O)(=O)O.[Fe]
153.917
Powder
250g
Fe 33 to 40% w/w
FeH2O4S
14,5094
MVZXTUSAYBWAAM-UHFFFAOYSA-N
iron;sulfuric acid
105075
Brown
Iron Dextran
Safety and Handling

Safety and Handling

GHS H Statement
H334-H351-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Suspected of causing cancer.
May cause an allergic skin reaction.

GHS P Statement
P284a-P261-P280-P342+P311-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
If experiencing respiratory symptoms: Call a POISON CENTER or doctor/physician.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

RTECSNumber : NI2200000