Learn More
Iron-chrome based high temperature gas shift catalyst, HiFUEL|r W210, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 04546736
Description
Iron-chrome based high temperature gas shift catalyst is ideal for use in fuel cell and other distributed hydrogen production applications. It is also used for fuel processing applications.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Iron-chrome based high temperature gas shift catalyst HiFUEL™ W210 | |
500 g | |
Pellets |
Red to Brown | |
Insoluble in water. |
Safety and Handling
GHS H Statement
H334-H340-H350-H361-H373-H332-H315-H319-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause genetic defects.
May cause cancer.
Suspected of damaging fertility or the unborn child.
May cause damage to organs through prolonged or repeated exposure.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.
P201-P202-P281-P308+P313-P501c
H340-H350
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only