missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Iron boride, 98%

Manufacturer:  Alfa Aesar™ 2295214

 View more versions of this product

Catalog No. AA2295214



Specifications

Specifications

Iron boride
BFe2
boron; iron, diiron boride, iron boride fe2b, boranylidyne diiron
[B].[Fe].[Fe]
122.5
122.5
−35 Mesh
25g
12006-85-8
MFCD00016092
FSDZRQFSRALZQJ-UHFFFAOYSA-N
boron;iron
6336853
98%
Odorless

Chemical Identifiers

12006-85-8
122.5
FSDZRQFSRALZQJ-UHFFFAOYSA-N
6336853
[B].[Fe].[Fe]
BFe2
MFCD00016092
boron; iron, diiron boride, iron boride fe2b, boranylidyne diiron
boron;iron
Safety and Handling

Safety and Handling

TSCA : TSCA

SDS Documents