Please login to your online account to display your discounted pricing

Alfa Aesar™ Iron boride, 98%

$152.19 - $152.19

Chemical Identifiers

Chemical Name or Material Iron boride
CAS 12006-85-8
Formula Weight 122.5
MDL Number MFCD00016092
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA2295222

  SDS
alfa aesar™
2295222
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA2295214

  SDS
alfa aesar™
2295214
25g Each for $152.19
Description & Specifications

Specifications

Formula Weight 122.5
Chemical Name or Material Iron boride
TSCA TSCA
Mesh Size −35 Mesh
Odor Odorless
Assay 98%
MDL Number MFCD00016092
CAS 12006-85-8
Quantity 100g