Please login to your online account to display your discounted pricing

Alfa Aesar™ Iron arsenide, 99.5% (metals basis)

Chemical Identifiers

Chemical Name or Material Iron arsenide
CAS 12005-88-8
Formula Weight 186.62
MDL Number MFCD00151533
UN Number UN1557
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA3619009

 
alfa aesar™
3619009
10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA3619018

 
alfa aesar™
3619018
50g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Lumps
Formula Weight 186.62
Chemical Name or Material Iron arsenide
TSCA TSCA
Odor Odorless
Particle Size 10mm & down
Assay 99.5%
Assay Percent Notes (metals basis)
MDL Number MFCD00151533
CAS 12005-88-8
UN Number UN1557
Quantity 10g
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H301-H331
Toxic if swallowed.
Toxic if inhaled.
Health Hazard 3 GHS P Statement
P261-P301+P310-P304+P340-P311-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POISON CENTER or doctor/physician.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: ARSENIC COMPOUNDS, SOLID, N.O.S.
Safety and Handling

Danger

GHS P Statement
P261-P301+P310-P304+P340-P311-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POISON CENTER or doctor/physician.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H331
Toxic if swallowed.
Toxic if inhaled.