Please login to your online account to display your discounted pricing

Alfa Aesar™ Iron aluminide, 97.5% (metals basis)

Chemical Identifiers

Chemical Name or Material Iron aluminide
CAS 12003-42-8
MDL Number MFCD00210617
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA4723318

 
alfa aesar™
4723318
50g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA4723330

 
alfa aesar™
4723330
250g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material Iron aluminide
Mesh Size −100 Mesh, Intermetallic
Physical Form Powder
Assay 97.5%
Assay Percent Notes (metals basis)
MDL Number MFCD00210617
CAS 12003-42-8
Color Gray
Melting Point 1540°C
Quantity 50g
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H319-H335
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Density 6.72g/mL
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H319-H335
Causes serious eye irritation.
May cause respiratory irritation.