missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Iron, Acros Organics™

Manufacturer:  Acros Organics AC449071000

 View more versions of this product

Catalog No. AC449071000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

Iron
Fe
Solubility in water: insoluble.
[Fe]
55.845
CHEBI:82664
ACS Reagent
0.02% max.
3000.0°C
100g
7439-89-6
ferrum, iron, elemental, ferrous, remko, armco, ferrovac, hoeganaes eh, 3zhp, ancor b, atomiron 5m
XEEYBQQBJWHFJM-UHFFFAOYSA-N
iron
23925
55.84
5ppm max.
Solubility in water: insoluble.
1535.0°C

Chemical Identifiers

7439-89-6
55.845
ferrum, iron, elemental, ferrous, remko, armco, ferrovac, hoeganaes eh, 3zhp, ancor b, atomiron 5m
CHEBI:82664
[Fe]
Fe
XEEYBQQBJWHFJM-UHFFFAOYSA-N
23925
iron
SDS Documents