Learn More
Iron AA Standard, Certified, 1000ppm ±5ppm (1mL = 1mg Fe), LabChem™
Supplier: LabChem LC157007
Specifications
Iron AA Standard | |
1000ppm (1mL = 1mg Fe) | |
Poly Bottle | |
96.42,2.86,0.72 | |
Fe | |
MFCD00010999 | |
XEEYBQQBJWHFJM-UHFFFAOYSA-N | |
iron | |
23925 | |
404.02 | |
Passes Test | |
Soluble in water |
1000ppm ±5ppm (1mL = 1mg Fe) | |
7439-89-6,7782-61-8,7782-61-8,7697-37-2,7732-18-5 | |
125 mL | |
Colorless | |
Fe(NO3)3·9H2O | |
UN3264 | |
[Fe] | |
55.85 | |
CHEBI:82664 | |
Certified | |
1g/mL | |
Liquid |
Chemical Identifiers
7439-89-6 | |
55.85 | |
XEEYBQQBJWHFJM-UHFFFAOYSA-N | |
CHEBI:82664 | |
[Fe] |
Fe | |
MFCD00010999 | |
23925 | |
iron |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
GHS P Statement
Do not breathe mist, vapors, spray.
Wear protective gloves/eye protection.
Wash exposed skin thoroughly after handling.
If swallowed: Rinse mouth.
Do not induce vomiting.
Immediately call a poison center/doctor.
If on skin (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Immediately call a poison center/doctor.
Wash contaminated clothing before reuse.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Immediately call a poison center/doctor.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a poison center/doctor.
Store locked up.
Dispose of contents/container to comply with local, state and federal regulations.
Danger
Recommended Storage : Room Temperature