Please login to your online account to display your discounted pricing

Iron AA, ICP and ICP-MS Standards, 10ppm Fe in 3 to 5% Nitric Acid, Ricca Chemical

Chemical Identifiers

Chemical Name or Material Iron AA, ICP and ICP-MS Standards
Name Note 10ppm Fe in 3 to 5% Nitric Acid
CAS 7439-89-6
CAS 7697-37-2
CAS 7732-18-5
Physical Form Liquid
Assay Percent Range 10.00 ± 0.05ppm
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

R4183500100

 
ricca chemical company
RIR4183500100A
Poly natural 100mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

R4183500500

 
ricca chemical company
RIR4183500500A
Poly natural 500mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Liquid
Color Clear, colorless to faint yellow
Quantity 100mL
Chemical Name or Material Iron AA, ICP and ICP-MS Standards
CAS 7439-89-6
Assay Percent Range 10.00 ± 0.05ppm
Solvent or Matrix Aqueous Solution
Name Note 10ppm Fe in 3 to 5% Nitric Acid
CAS 7732-18-5
CAS 7697-37-2
Solubility Solubility in Water: Infinite
Odor Odorless
Packaging Poly natural