missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Iron, 99%, powder, -70 mesh (<212 micron), ACROS Organics™

Manufacturer:  Acros Organics AC197810100

 View more versions of this product

Catalog No. AC197810100

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Reduction reagent
Specifications

Specifications

Iron
7439-89-6
7.8600g/mL
Fe
01,519; 02,229; 03,167; 05,357; 09,257; 14,184; 16,189; 17,157
ferrum, iron, elemental, ferrous, remko, armco, ferrovac, hoeganaes eh, 3zhp, ancor b, atomiron 5m
500ppm max.
[Fe]
55.845
23925
55.84
99%
50ppm max.
1535.0°C
10kg
100ppm max.
99%
MFCD00010999
15, 5137
1000ppm max.
XEEYBQQBJWHFJM-UHFFFAOYSA-N
iron
Plastic bottle
CHEBI:82664
Fine Powder
200ppm max.
Gray
3000.0°C

Chemical Identifiers

7439-89-6
55.845
XEEYBQQBJWHFJM-UHFFFAOYSA-N
23925
iron
Fe
MFCD00010999
ferrum, iron, elemental, ferrous, remko, armco, ferrovac, hoeganaes eh, 3zhp, ancor b, atomiron 5m
CHEBI:82664
[Fe]
SDS Documents