missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Iron, 99%, powder, -70 mesh (<212 micron), ACROS Organics™

$37.51 - $244.86

Chemical Identifiers

CAS 7439-89-6
Physical Form Fine Powder
Assay Percent Range 99%
Molecular Formula Fe
Formula Weight 55.84
MDL Number MFCD00010999
Chemical Name or Material Iron
Assay ≥98.5% (Fe)
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
AC197810100 SDS Thermo Fisher Scientific
AC197810100
Plastic bottle 10kg N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC197811000 SDS Thermo Fisher Scientific
AC197811000
Plastic bottle 100g Each for $37.51
AC197810020 SDS Thermo Fisher Scientific
197810020
Plastic bottle 2kg Each for $244.86
AC197815000 SDS Thermo Fisher Scientific
197815000
Plastic bottle 500g Each for $95.86
Specifications

Description

Reduction reagent
Description & Specifications

Chemical Identifiers

7439-89-6
99%
55.84
Iron
Fine Powder
Fe
MFCD00010999
≥98.5% (Fe)

Specifications

Fine Powder
10kg
7.8600g/mL
15, 5137
1000ppm max.
Fe
7439-89-6
Plastic bottle
200ppm max.
≥98.5% (Fe)
55.84
Gray
01,519; 02,229; 03,167; 05,357; 09,257; 14,184; 16,189; 17,157
50ppm max.
100ppm max.
MFCD00010999
99%
500ppm max.
Iron