missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Iron, 99.99%, (trace metal basis), granules, ACROS Organics™

Manufacturer:  Acros Organics AC437110050

 View more versions of this product

Catalog No. AC437110050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

For ultra trace analysis, Reduction reagent
Specifications

Specifications

Iron
7439-89-6
Fe
XEEYBQQBJWHFJM-UHFFFAOYSA-N
iron
Glass bottle
CHEBI:82664
Granules
Trace Metal Basis
Undesignated
3000.0°C
5g
99.99%
ferrum, iron, elemental, ferrous, remko, armco, ferrovac, hoeganaes eh, 3zhp, ancor b, atomiron 5m
[Fe]
55.845
23925
55.84
99.99%
50ppm max.
1535.0°C

Chemical Identifiers

7439-89-6
55.845
ferrum, iron, elemental, ferrous, remko, armco, ferrovac, hoeganaes eh, 3zhp, ancor b, atomiron 5m
CHEBI:82664
[Fe]
Fe
XEEYBQQBJWHFJM-UHFFFAOYSA-N
23925
iron
SDS Documents