Please login to your online account to display your discounted pricing

Iron, 99.99%, (trace metal basis), granules, ACROS Organics™

$176.80 - $176.80

Chemical Identifiers

Chemical Name or Material Iron
CAS 7439-89-6
Purity Grade Trace Metal Basis
Assay Percent Range 99.99%
Molecular Formula Fe
Formula Weight 55.84
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC437110050

  SDS
thermo fisher scientific
AC437110050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC437110250

  SDS
thermo fisher scientific
AC437110250
Glass bottle 25g Each for $176.80
Description & Specifications

Specifications

Purity Grade Trace Metal Basis
Physical Form Granules
Formula Weight 55.84
Color Silvery
Quantity 5g
Chemical Name or Material Iron
CAS 7439-89-6
Assay Percent Range 99.99%
Molecular Formula Fe
Packaging Glass bottle
Total Trace Metal Impurities 50ppm max.

For ultra trace analysis, Reduction reagent