missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Iron

$12.70 - $12.70

Chemical Identifiers

CAS 7439-89-6
Linear Formula Fe
Formula Weight 55.85
Chemical Name or Material Iron
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
S96863 Fisher Science Education
IX0210-500G
Metal Mesh (40-60 Mesh) 500g Each for $12.70
Specifications Description & Specifications

Chemical Identifiers

7439-89-6
Fe
55.85
Iron

Specifications

500g
Fe
Metal Mesh (40-60 Mesh)
Gray-black
7439-89-6
55.85
Certificates of Analysis are not available for Fisher Scientific™ education-grade chemicals. A hazardous surcharge will be applied to chemicals that are designated Hazardous by the U.S. Dept. of Transportation at the time of shipment.
Iron

Safety and Handling

ShelfLife : Indefinite

DOTInformation : DOT Class 4.1, : Flammable Solid