Please login to your online account to display your discounted pricing

Iron(0) pentacarbonyl, AcroSeal™, ACROS Organics™

$168.35 - $168.35

Chemical Identifiers

Chemical Name or Material Iron(0) Pentacarbonyl
CAS 13463-40-6
Molecular Formula C5FeO5
Linear Formula Fe(CO)5
Formula Weight 195.9
Synonym Pentacarbonyl Iron
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC187621000

  SDS
thermo fisher scientific
AC187621000
AcroSeal™ Glass Bottle in Tin Can 100mL Each for $168.35

AC187620010

  SDS
thermo fisher scientific
AC187620010
AcroSeal™ Glass Bottle 1L CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Liquid Which May Contain Particulate Matter
Formula Weight 195.9
Quantity 100mL
Color Yellow-orange to brown
UN Number 1994
Packaging AcroSeal™ Glass Bottle in Tin Can
Density 1.4900g/mL
Chemical Name or Material Iron(0) pentacarbonyl
Molecular Formula C5FeO5
Linear Formula Fe(CO)5
CAS 13463-40-6
Synonym Pentacarbonyl iron
Health Hazard 1 Danger
Additional Information Material may have to be filtered through glass wool before use. Keep residue moist. May be pyrophoric if allowed to dry
Health Hazard 2 GHS H Statement:
Fatal in contact with skin.
Fatal if inhaled.
Highly flammable liquid and vapour.
Fatal if swallowed.
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Infrared Spectrum Authentic
Safety and Handling

Danger

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Fatal in contact with skin.
Fatal if inhaled.
Highly flammable liquid and vapour.
Fatal if swallowed.