Please login to your online account to display your discounted pricing

Alfa Aesar™ IRL-1038

For cellular and molecular biology applications

Specifications

Product Type IRL-1038
Molecular Weight 1409.67
Storage Requirements -30° to -10°C
Sequence Cys-Val-Tyr-Phe-Cys-His-Leu-Asp-Ile-Ile-Trp (Disulfide bridge Cys1-Cys5)
Format Solid
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ66476EXD

 
alfa aesar™
J66476EXD
0.5mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAJ66476MCR

 
alfa aesar™
J66476MCR
1mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Product Type IRL-1038
Molecular Weight 1409.67
Storage Requirements -30° to -10°C
Sequence Cys-Val-Tyr-Phe-Cys-His-Leu-Asp-Ile-Ile-Trp (Disulfide bridge Cys1-Cys5)
Format Solid
Common Name Endothelin β Receptor Antagonist IRL 1038
Regulatory Status RUO
Purification Method HPLC