Please login to your online account to display your discounted pricing

Alfa Aesar™ Iridium wire, 1.0mm (0.04 in.) dia., 99.8% (metals basis)

Chemical Identifiers

Chemical Name or Material Iridium wire
CAS 7439-88-5
Name Note 1.0mm (0.04 in.) dia.
MDL Number MFCD00011062
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA11429BO

  SDS
alfa aesar™
11429BO
1cm CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA11429BQ

  SDS
alfa aesar™
11429BQ
5cm CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material Iridium wire
TSCA TSCA
Name Note 1.0mm (0.04 in.) dia.
Physical Form Wire
Assay 99.8%
Assay Percent Notes (metals basis)
MDL Number MFCD00011062
CAS 7439-88-5
Weight ≈17.8g/m
Quantity 1cm