missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Iridium wire, 0.5mm (0.02 in.) dia., 99.8% (metals basis)

$279.08 - $1,040.80

Chemical Identifiers

CAS 7439-88-5
Molecular Formula Ir
Molecular Weight (g/mol) 192.217
MDL Number MFCD00011062
InChI Key GKOZUEZYRPOHIO-UHFFFAOYSA-N
Synonym atom, black, hydrido-iridium, ion, iridio, iridium, elemental, iridium, ion ir4, metallicum, powder, sponge
PubChem CID 23924
ChEBI CHEBI:49666
IUPAC Name iridium
SMILES [Ir]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA11430BQ
View Documents
Alfa Aesar™
11430BQ
5cm Each for $279.08
 
AA11430BU
View Documents
Alfa Aesar™
11430BU
25cm Each for $1,040.80
 
Specifications

Chemical Identifiers

7439-88-5
192.217
GKOZUEZYRPOHIO-UHFFFAOYSA-N
23924
iridium
Ir
MFCD00011062
atom, black, hydrido-iridium, ion, iridio, iridium, elemental, iridium, ion ir4, metallicum, powder, sponge
CHEBI:49666
[Ir]

Specifications

≈4.45g/m
7439-88-5
MFCD00011062
GKOZUEZYRPOHIO-UHFFFAOYSA-N
iridium
23924
Wire, ≈4.45g/m
Iridium wire, 0.5mm (0.02 in.) dia.
5cm
Ir
atom, black, hydrido-iridium, ion, iridio, iridium, elemental, iridium, ion ir4, metallicum, powder, sponge
[Ir]
192.217
CHEBI:49666
99.8%
(metals basis)
Safety and Handling

Safety and Handling

EINECSNumber : 231-095-9

TSCA : Yes

Recommended Storage : Ambient temperatures