missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Iridium wire, 0.5mm (0.02 in.) dia., 99.8% (metals basis)

Manufacturer:  Alfa Aesar™ 11430BU

 View more versions of this product

Catalog No. AA11430BU



Specifications

Specifications

Iridium wire
25cm
7439-88-5
Ir
atom, black, hydrido-iridium, ion, iridio, iridium, elemental, iridium, ion ir4, metallicum, powder, sponge
[Ir]
192.217
CHEBI:49666
99.8%
≈4.45g/m
0.5mm (0.02 in.) dia.
(metals basis)
MFCD00011062
GKOZUEZYRPOHIO-UHFFFAOYSA-N
iridium
23924
Wire, ≈4.45g/m

Chemical Identifiers

7439-88-5
192.217
GKOZUEZYRPOHIO-UHFFFAOYSA-N
23924
iridium
Ir
MFCD00011062
atom, black, hydrido-iridium, ion, iridio, iridium, elemental, iridium, ion ir4, metallicum, powder, sponge
CHEBI:49666
[Ir]
Safety and Handling

Safety and Handling

EINECSNumber : 231-095-9

TSCA : Yes

Recommended Storage : Ambient temperatures

Documents