missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Iridium Wire, 0.25mm (0.01 in.) dia., 99.8% (Metals basis), Alfa Aesar™

Form: Approx.1.11g/m

Manufacturer:  Thermo Fisher Scientific 11432CA

 View more versions of this product

Catalog No. AA11432CA



Specifications

Specifications

Iridium
7439-88-5
22.65g/cm3 at 20°C
Ir
atom, black, hydrido-iridium, ion, iridio, iridium, elemental, iridium, ion ir4, metallicum, powder, sponge
[Ir]
192.217
CHEBI:49666
White to Yellow
200cm
White to pale yellow wire
99.8% (Metals basis)
MFCD00011062
GKOZUEZYRPOHIO-UHFFFAOYSA-N
iridium
23924
Wire, ≈1.11g/m

Chemical Identifiers

7439-88-5
192.217
GKOZUEZYRPOHIO-UHFFFAOYSA-N
23924
iridium
Ir
MFCD00011062
atom, black, hydrido-iridium, ion, iridio, iridium, elemental, iridium, ion ir4, metallicum, powder, sponge
CHEBI:49666
[Ir]
Safety and Handling

Safety and Handling

EINECSNumber : 231-095-9

TSCA : Yes

Recommended Storage : Ambient temperatures

Documents