missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Iridium wire, 0.075mm (0.003 in.) dia., 99.9% (metals basis)

Manufacturer:  Alfa Aesar™ 44714BW

 View more versions of this product

Catalog No. AA44714BW



Specifications

Specifications

Iridium wire
50cm
7439-88-5
Ir
ion, black, metallicum, iridium, elemental, iridium, ion ir4, iridio, hydrido-iridium, atom, powder, sponge
[Ir]
192.217
CHEBI:49666
99.9%
≈0.099g/m
0.075mm (0.003 in.) dia.
(metals basis)
MFCD00011062
GKOZUEZYRPOHIO-UHFFFAOYSA-N
iridium
23924
Wire

Chemical Identifiers

7439-88-5
192.217
GKOZUEZYRPOHIO-UHFFFAOYSA-N
23924
iridium
Ir
MFCD00011062
ion, black, metallicum, iridium, elemental, iridium, ion ir4, iridio, hydrido-iridium, atom, powder, sponge
CHEBI:49666
[Ir]
Safety and Handling

Safety and Handling

TSCA : TSCA

SDS Documents