missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Iridium slug, 6.0mm (0.24 in.) dia. x 6.0mm (0.24 in.) length, 99.8% (metals basis)

Manufacturer:  Alfa Aesar™ 44232KG

 View more versions of this product

Catalog No. AA44232KG



Specifications

Specifications

Iridium slug
1 pc.
7439-88-5
Ir
atom
[Ir]
192.217
CHEBI:49666
99.8%
≈4.85g/slug
6.0mm (0.24 in.) dia. x 6.0mm (0.24 in.) length
(metals basis)
MFCD00011062
GKOZUEZYRPOHIO-UHFFFAOYSA-N
iridium
23924
≈4.85g/slug

Chemical Identifiers

7439-88-5
192.217
GKOZUEZYRPOHIO-UHFFFAOYSA-N
23924
iridium
Ir
MFCD00011062
atom
CHEBI:49666
[Ir]
Safety and Handling

Safety and Handling

EINECSNumber : 231-095-9

TSCA : Yes

Recommended Storage : Ambient temperatures

Documents