Please login to your online account to display your discounted pricing

Alfa Aesar™ Iridium powder, -60 mesh, 99.95% (metals basis)

Chemical Identifiers

Chemical Name or Material Iridium powder
CAS 7439-88-5
MDL Number MFCD00011062
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA1207002

  SDS
alfa aesar™
1207002
0.5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA1207009

  SDS
alfa aesar™
1207009
10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA1207004

  SDS
alfa aesar™
1207004
2g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Quantity 0.5g
Mesh Size −60 Mesh
CAS 7439-88-5
Chemical Name or Material Iridium powder
MDL Number MFCD00011062
Physical Form Powder
Assay Percent Notes (metals basis)
Color Gray
Assay 99.95%
TSCA TSCA