Please login to your online account to display your discounted pricing

Alfa Aesar™ Iridium powder, -325 mesh, 99.9% (metals basis)

$361.73 - $361.73

Chemical Identifiers

Chemical Name or Material Iridium powder
CAS 7439-88-5
MDL Number MFCD00011062
UN Number UN3089
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA3968404

 
alfa aesar™
3968404
2g Each for $361.73

AA3968402

 
alfa aesar™
3968402
0.5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Health Hazard 3 GHS P Statement
P210-P280-P240-P241-P370+P378a
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Ground/bond container and receiving equipment.
Use explosion-proof electrical/ventilating/lighting/ equipment.
In case of fire: Use for extinction: CO2, powder or water spray.
Chemical Name or Material Iridium powder
TSCA TSCA
Mesh Size −325 Mesh
Physical Form Powder/Sponge
Assay 99.9%
Assay Percent Notes (metals basis)
MDL Number MFCD00011062
CAS 7439-88-5
UN Number UN3089
Quantity 2g
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H228
Flammable solid.
DOT Information Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: METAL POWDERS, FLAMMABLE, N.O.S.
Safety and Handling

Warning

GHS P Statement
P210-P280-P240-P241-P370+P378a
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Ground/bond container and receiving equipment.
Use explosion-proof electrical/ventilating/lighting/ equipment.
In case of fire: Use for extinction: CO2, powder or water spray.

GHS H Statement
H228
Flammable solid.