Please login to your online account to display your discounted pricing

Alfa Aesar™ Iridium powder, -22 mesh, Premion™, 99.99% (metals basis)

Chemical Identifiers

Chemical Name or Material Iridium powder
CAS 7439-88-5
Purity Grade Premion™
MDL Number MFCD00011062
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA1074603

  SDS
alfa aesar™
1074603
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA1074606

  SDS
alfa aesar™
1074606
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material Iridium powder
TSCA TSCA
Mesh Size −22 Mesh
Purity Grade Premion™
Physical Form Powder
Assay 99.99%
Assay Percent Notes (metals basis)
MDL Number MFCD00011062
CAS 7439-88-5
Color Gray
Quantity 1g