Please login to your online account to display your discounted pricing

Iridium plasma standard solution, Ir 10,000μg/mL, Alfa Aesar™, Specpure™

Chemical Identifiers

Chemical Name or Material Iridium
Physical Form Liquid
Molecular Formula IrCl3 in 20% HCl
MDL Number MFCD00011062
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA35752AP

  SDS
alfa aesar
35752AP
500mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA35752AD

  SDS
alfa aesar
35752AD
50mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA35752AE

  SDS
alfa aesar
35752AE
100mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Quantity 500mL
DOT Information Hazard Class: 8, Packing Group: II
Density Approx. 1g/cm3
Physical Form Liquid
Molecular Formula IrCl3 in 20% HCl
EINECS Number 231-595-7
Chemical Name or Material Iridium
MDL Number MFCD00011062
Health Hazard 1 H314-H335: Causes severe skin burns and eye damage. May cause respiratory irritation.
Odor Characteristic
Appearance Liquid
Safety and Handling

H314-H335: Causes severe skin burns and eye damage. May cause respiratory irritation.