missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Iridium(IV) oxide, Premion™, 99.99% (metals basis), Ir 84.5% min

Manufacturer:  Alfa Aesar™ 4339606

 View more versions of this product

Catalog No. AA4339606



Specifications

Specifications

Iridium (IV) oxide
(metals basis)
MFCD00011065
224.2
99.99%
Ir 84.5% min
Odorless
5g
12030-49-8
IrO2
UN1479
Powder
Premion™
11.7g/mL
Black

Chemical Identifiers

12030-49-8
IrO2
MFCD00011065
Safety and Handling

Safety and Handling

GHS H Statement
H272
May intensify fire
oxidizer.

GHS P Statement
P221-P210-P220-P280-P370+P378a-P501a
Take any precaution to avoid mixing with combustibles.
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Keep/Store away from clothing/combustible materials.
Wear protective gloves/protective clothing/eye protection/face protection.
In case of fire: Use for extinction: CO2, powder or water spray.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 5.1; Packing Group: II; Proper Shipping Name: OXIDIZING SOLID, N.O.S.

TSCA : TSCA

Documents