missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Iridium(IV) oxide, Premion™, 99.99% (metals basis), Ir 84.5% min

Manufacturer:  Alfa Aesar™ 4339603

 View more versions of this product

Catalog No. AA4339603



Description

Description

Specifications

Specifications

Iridium (IV) oxide
(metals basis)
MFCD00011065
Insoluble in water.
Powder
Premion™
11.7 g/mL
Black
12030-49-8
IrO2
UN1479
224.2
99.99%
Ir 84.5% min
Odorless
1g

Chemical Identifiers

12030-49-8
IrO2
MFCD00011065
Safety and Handling

Safety and Handling

GHS H Statement
H272
May intensify fire
oxidizer.

P210-P220-P221-P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362-P370+P378q-P501c

H272-H315-H319

DOTInformation : Transport Hazard Class: 5.1; Packing Group: II; Proper Shipping Name: OXIDIZING SOLID, N.O.S.

EINECSNumber : 234-743-9

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents