Please login to your online account to display your discounted pricing

Alfa Aesar™ Iridium(IV) oxide powder, 99%

Chemical Identifiers

Chemical Name or Material Iridium(IV) oxide powder
CAS 12030-49-8
Molecular Formula IrO2
Formula Weight 224.2
MDL Number MFCD00011065
UN Number UN1479
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1784906

  SDS
alfa aesar™
A1784906
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAA1784903

  SDS
alfa aesar™
A1784903
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material Iridium(IV) oxide powder
CAS 12030-49-8
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H272
May intensify fire; oxidizer.
Health Hazard 3 GHS P Statement
P221-P210-P220-P280-P370+P378a-P501a
Take any precaution to avoid mixing with combustibles.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Keep/Store away from clothing/combustible materials.
Wear protective gloves/protective clothing/eye protection/face protection.
In case of fire: Use for extinction: CO2, powder or water spray.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 5.1; Packing Group: II; Proper Shipping Name: OXIDIZING SOLID, N.O.S.
TSCA TSCA
Assay 99%
Molecular Formula IrO2
Formula Weight 224.2
MDL Number MFCD00011065
UN Number UN1479
Odor Odorless
Quantity 5g

Applications
Iridium(IV) oxide is used in the coating of anode-electrodes for industrial electrolysis in association with other rare oxides. It is also used in microelectrodes for electrophysiology research.

Solubility
Insoluble in water.

Notes
Incompatible with strong oxidizing agents.

Safety and Handling

Danger

GHS P Statement
P221-P210-P220-P280-P370+P378a-P501a
Take any precaution to avoid mixing with combustibles.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Keep/Store away from clothing/combustible materials.
Wear protective gloves/protective clothing/eye protection/face protection.
In case of fire: Use for extinction: CO2, powder or water spray.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H272
May intensify fire; oxidizer.