Please login to your online account to display your discounted pricing

Alfa Aesar™ Iridium(IV) iodide, Premion™, 99.95% (metals basis), Ir 27.0% min

Chemical Identifiers

Chemical Name or Material Iridium(IV) iodide
CAS 7790-45-6
Purity Grade Premion™
Formula Weight 699.82
MDL Number MFCD00049960
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA4042403

  SDS
alfa aesar™
4042403
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA4042406

  SDS
alfa aesar™
4042406
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 699.82
Assay Percent Notes (metals basis)
Chemical Name or Material Iridium(IV) iodide
Assay 99.95%
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Purity Grade Premion™
Quantity 1g
MDL Number MFCD00049960
Health Hazard 1 Warning
CAS 7790-45-6
Concentration Ir 27.0% min.
Physical Form Powder
TSCA TSCA
Solubility Information Insoluble in water and alcohol; Soluble in aqueous KI
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.