missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Iridium(IV) chloride, 99.95% (metals basis), Ir 56.5% min., Alfa Aesar™, Premion™

Manufacturer:  Alfa Aesar 1218403

 View more versions of this product

Catalog No. AA1218403



Specifications

Specifications

Iridium(IV) chloride
99.95% (metals basis)
MFCD00016155
CALMYRPSSNRCFD-UHFFFAOYSA-J
tetrachloroiridium
24815
Glassy Amorphous Pieces/Powder
10025-97-5
Cl4Ir
iridium iv chloride, iridium tetrachloride, iridium chloride ircl4, iridic chloride, unii-pcg7kvc21i, pcg7kvc21i, iridium iv chloride, iridium chloride ircl4, acmc-1cp8w, iridium iv chloride, premion
Cl[Ir](Cl)(Cl)Cl
334.017
334.01
1g

Chemical Identifiers

10025-97-5
334.017
CALMYRPSSNRCFD-UHFFFAOYSA-J
24815
Cl[Ir](Cl)(Cl)Cl
Cl4Ir
MFCD00016155
iridium iv chloride, iridium tetrachloride, iridium chloride ircl4, iridic chloride, unii-pcg7kvc21i, pcg7kvc21i, iridium iv chloride, iridium chloride ircl4, acmc-1cp8w, iridium iv chloride, premion
tetrachloroiridium
Safety and Handling

Safety and Handling

H302: Harmful if swallowed.

EINECSNumber : 233-048-8

Documents