Please login to your online account to display your discounted pricing

Iridium(IV) chloride, 99.95% (metals basis), Ir 56.5% min., Alfa Aesar™, Premion™

Chemical Identifiers

Chemical Name or Material Iridium(IV) chloride
CAS 10025-97-5
Physical Form Glassy Amorphous Pieces/Powder
Assay Percent Range 99.95% (Metal basis)
Molecular Formula IrCl4
Formula Weight 334.01
MDL Number MFCD00016155
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA1218401

  SDS
alfa aesar™
1218401
0.25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA1218403

  SDS
alfa aesar
1218403
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 334.01
Physical Form Glassy Amorphous Pieces/Powder
Quantity 0.25g
CAS 10025-97-5
EINECS Number 233-048-8
Molecular Formula IrCl4
Appearance Drak brown to black glassy amorphous pieces/powder
MDL Number MFCD00016155
Chemical Name or Material Iridium(IV) chloride
Assay Percent Range 99.95% (Metal basis)
Health Hazard 1 H302: Harmful if swallowed.
Safety and Handling

H302: Harmful if swallowed.