missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Iridium(III) chloride trihydrate, 53-56% Ir, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 195500250
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Iridium(III) chloride trihydrate | |
Black to Green | |
Crystals | |
Cl3Ir·3H2O | |
01,132; 02,67 | |
iridium iii chloride trihydrate, trichloroiridium trihydrate, cl3ir.3h2o, acmc-1c5tf, iridium chloride ircl3 , trihydrate 7ci,8ci,9ci | |
LNJXVUXPFZKMNF-UHFFFAOYSA-K | |
trichloroiridium;trihydrate | |
6102153 | |
53 to 56% Ir | |
5.3 |
763.0°C | |
13569-57-8 | |
25 g | |
IrCl3·3H2O | |
14, 5088 | |
Solubility in water: insoluble in water. Other solubilities: insoluble in acids ans alkalies | |
O.O.O.Cl[Ir](Cl)Cl | |
352.6 | |
352.6 | |
Glass bottle | |
5.3000g/mL |
Chemical Identifiers
13569-57-8 | |
352.6 | |
iridium iii chloride trihydrate, trichloroiridium trihydrate, cl3ir.3h2o, acmc-1c5tf, iridium chloride ircl3 , trihydrate 7ci,8ci,9ci | |
trichloroiridium;trihydrate |
Cl3Ir·3H2O | |
LNJXVUXPFZKMNF-UHFFFAOYSA-K | |
6102153 | |
O.O.O.Cl[Ir](Cl)Cl |
RUO – Research Use Only