missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Iridium(III) chloride hydrate, Black crystalline, 99.9% (Metals basis), Alfa Aesar™

$170.36 - $2,875.28

Chemical Identifiers

CAS 14996-61-3
Molecular Formula Cl3H2IrO
Molecular Weight (g/mol) 316.58
MDL Number MFCD00149750
InChI Key MJRFDVWKTFJAPF-UHFFFAOYSA-K
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA1103003
View Documents
Thermo Fisher Scientific
1103003
1g
Each for $170.36
 
AA1103006
View Documents
Thermo Fisher Scientific
1103006
5g
Each for $621.99
 
AA1103014
View Documents
Thermo Fisher Scientific
1103014
25g
Each for $2,875.28
 
Description

Description

Specifications

Chemical Identifiers

14996-61-3
316.58
MJRFDVWKTFJAPF-UHFFFAOYSA-K
Cl3H2IrO
MFCD00149750

Specifications

14996-61-3
Cl3H2IrO
Soluble in water and alcohol.
316.58
Crystals
Black
Iridium(III) chloride hydrate
99.9% (Metals basis)
MFCD00149750
MJRFDVWKTFJAPF-UHFFFAOYSA-K
298.56 (Anhydrous)
Odorless
1g
Safety and Handling

Safety and Handling

P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c

H314-H335

DOTInformation : Hazard Class: 8; Packaging Group: II

EINECSNumber : 233-044-6

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS