missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Iridium(III) chloride, Anhydrous, Ir 62% min., Alfa Aesar™
Manufacturer: Alfa Aesar 1215803
Specifications
Iridium(IV) chloride | |
99.95% (Metal basis) | |
MFCD00016155 | |
iridium iv chloride, iridium tetrachloride, iridium chloride ircl4, iridic chloride, unii-pcg7kvc21i, pcg7kvc21i, iridium iv chloride, iridium chloride ircl4, acmc-1cp8w, iridium iv chloride, premion | |
Cl[Ir](Cl)(Cl)Cl | |
334.017 | |
334.01 | |
Black to Brown |
10025-97-5 | |
Cl4Ir | |
Drak brown to black glassy amorphous pieces/powder | |
CALMYRPSSNRCFD-UHFFFAOYSA-J | |
tetrachloroiridium | |
24815 | |
Glassy Amorphous Pieces/Powder | |
1g |
Chemical Identifiers
10025-97-5 | |
334.017 | |
CALMYRPSSNRCFD-UHFFFAOYSA-J | |
24815 | |
Cl[Ir](Cl)(Cl)Cl |
Cl4Ir | |
MFCD00016155 | |
iridium iv chloride, iridium tetrachloride, iridium chloride ircl4, iridic chloride, unii-pcg7kvc21i, pcg7kvc21i, iridium iv chloride, iridium chloride ircl4, acmc-1cp8w, iridium iv chloride, premion | |
tetrachloroiridium |
Safety and Handling
H302: Harmful if swallowed.
EINECSNumber : 233-048-8