Please login to your online account to display your discounted pricing

Alfa Aesar™ Iridium(III) bromide hydrate, Ir 35.9% min

Chemical Identifiers

Chemical Name or Material Iridium(III) bromide hydrate
Formula Weight 431.91 (Anhydrous)
MDL Number MFCD00049466
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA1221203

 
alfa aesar™
1221203
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 431.91 (Anhydrous)
Melting Point 100°C (in -3H2O)
Chemical Name or Material Iridium(III) bromide hydrate
TSCA TSCA
Physical Form Crystalline
MDL Number MFCD00049466
Quantity 1g
Concentration Ir 35.9% min.