Please login to your online account to display your discounted pricing

Alfa Aesar™ Iridium(III) 2,4-pentanedionate, Ir 37.5% min

Chemical Identifiers

Chemical Name or Material Iridium(III) 2,4-pentanedionate
CAS 15635-87-7
Formula Weight 489.53
MDL Number MFCD00015353
Synonym Iridium(III) acetylacetonate
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA1051403

  SDS
alfa aesar™
1051403
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA1051401

  SDS
alfa aesar™
1051401
0.25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA1051406

  SDS
alfa aesar™
1051406
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 489.53
Physical Form Crystalline
MDL Number MFCD00015353
CAS 15635-87-7
Synonym Iridium(III) acetylacetonate
Melting Point 269° to 271°C
Quantity 1g
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Concentration Ir 37.5% min.
Chemical Name or Material Iridium(III) 2,4-pentanedionate
Odor Odorless
Color Orange-yellow
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.