missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Iridium(III) 2,4-pentanedionate, Ir 37.5% min

Manufacturer:  Alfa Aesar™ 1051401

 View more versions of this product

Catalog No. AA1051401



Specifications

Specifications

Iridium(III) 2,4-pentanedionate
C15H21IrO6-3
Iridium(III) acetylacetonate
CC(=CC(=O)C)[O-].CC(=CC(=O)C)[O-].CC(=CC(=O)C)[O-].[Ir]
489.544
489.53
Ir 37.5% min.
Orange-Yellow
0.25g
15635-87-7
MFCD00015353
AZFHXIBNMPIGOD-LNTINUHCSA-K
iridium;(Z)-4-oxopent-2-en-2-olate
117590592
Crystalline
Odorless
269°C to 271°C

Chemical Identifiers

15635-87-7
489.544
AZFHXIBNMPIGOD-LNTINUHCSA-K
117590592
CC(=CC(=O)C)[O-].CC(=CC(=O)C)[O-].CC(=CC(=O)C)[O-].[Ir]
C15H21IrO6-3
MFCD00015353
Iridium(III) acetylacetonate
iridium;(Z)-4-oxopent-2-en-2-olate
Safety and Handling

Safety and Handling

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

Documents