Please login to your online account to display your discounted pricing

Alfa Aesar™ Iridium foil, 1.0mm (0.04 in.) thick, typically 99.7+% (metals basis)

Chemical Identifiers

Chemical Name or Material Iridium foil
CAS 7439-88-5
Name Note 1.0mm (0.04 in.) thick
MDL Number MFCD00011062
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA11525FF

  SDS
alfa aesar™
11525FF
25x25mm CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥99.7% (typically)
Chemical Name or Material Iridium foil
TSCA TSCA
Name Note 1.0mm (0.04 in.) thick
Physical Form Foil
Assay Percent Notes (metals basis)
MDL Number MFCD00011062
CAS 7439-88-5
Weight ≈14.2g/25x25mm
Quantity 25x25mm